Please login to your online account to display your discounted pricing

4-Methylpyridazine, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  305830010

 View more versions of this product

Catalog No. AC305830010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 1.0600g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5170 to 1.5200 (20°C, 589nm)
Color Clear yellow
Quantity 1g
Chemical Name or Material 4-Methylpyridazine
Name Note 97%
CAS 1120-88-3
CAS Min % 96.0
CAS Max % 100.0
Physical Form Clear yellow liquid
Assay 0.96
Assay Percent Range 97%
Molecular Formula C5H6N2
Formula Weight 94.12g/mol
MDL Number MFCD00011594
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Methylpyridazine
Name Note 97%
CAS 1120-88-3
CAS Min % 96.0
CAS Max % 100.0
Assay 0.96
Assay Percent Range 97%
Molecular Formula C5H6N2
Formula Weight 94.12g/mol
MDL Number MFCD00011594
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents