Please login to your online account to display your discounted pricing

4-Methylpyridazine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methylpyridazine
Name Note 97%
CAS 1120-88-3
CAS Min % 96.0
CAS Max % 100.0
Assay 0.96
Assay Percent Range 97%
Molecular Formula C5H6N2
Formula Weight 94.12g/mol
MDL Number MFCD00011594
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305830010

  SDS
thermo fisher scientific
305830010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 1.0600g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5170 to 1.5200 (20°C, 589nm)
Color Clear yellow
Quantity 1g
Chemical Name or Material 4-Methylpyridazine
Name Note 97%
CAS 1120-88-3
CAS Min % 96.0
CAS Max % 100.0
Physical Form Clear yellow liquid
Assay 0.96
Assay Percent Range 97%
Molecular Formula C5H6N2
Formula Weight 94.12g/mol
MDL Number MFCD00011594
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.