Learn More
4-Methylpyrazole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 206970010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Methylpyrazole | |
0.9900g/mL | |
96°C | |
96% min. (GC) | |
C4H6N2 | |
1g | |
0.99 | |
4-methylpyrazole, fomepizole, antizol, 1h-pyrazole, 4-methyl, fomepizol, 4-methylpyrazol, fomepizolum, fomepizol inn-spanish, fomepizolum inn-latin, fomepizole usan:inn | |
RIKMMFOAQPJVMX-UHFFFAOYSA-N | |
4-methyl-1H-pyrazole | |
3406 | |
82.11 |
7554-65-6 | |
99.0°C to 100.0°C (6.0mmHg) | |
Authentic | |
Glass bottle | |
1.4930 to 1.4950 | |
23,65 | |
15,4253 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
CC1=CNN=C1 | |
82.11 | |
CHEBI:5141 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 231-445-
RUO â Research Use Only