Please login to your online account to display your discounted pricing

4-Methylpyrazole, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC206970250

 View more versions of this product

Catalog No. AC206970250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid After Melting
Assay ≥96.0% (GC)
Formula Weight 82.11
Quantity 25g
Color Clear colorless to yellow
Density 0.9900g/mL
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Chemical Name or Material 4-Methylpyrazole
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C4H6N2
CAS 7554-65-6
Health Hazard 1 Warning

Specifications

Chemical Name or Material 4-Methylpyrazole
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C4H6N2
Formula Weight 82.11
Accessories

Warning

SDS & Safety Documents