Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Methylpropiophenone, 94%

$38.18 - $143.61

Chemical Identifiers

Chemical Name or Material 4'-Methylpropiophenone
CAS 5337-93-9
Molecular Formula C10H12O
Formula Weight 148.21
MDL Number MFCD00009312
Beilstein 2042137
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2167322

 
alfa aesar™
B2167322
100g Each for $38.18

AAB2167336

 
alfa aesar™
B2167336
500g Each for $143.61
Description & Specifications

Specifications

Formula Weight 148.21
Chemical Name or Material 4'-Methylpropiophenone
Assay 94%
Molecular Formula C10H12O
MDL Number MFCD00009312
CAS 5337-93-9
Beilstein 2042137
Melting Point 7°C
Boiling Point 238° to 239°C
Flash Point 108°C (226°F)
Quantity 100g

Applications
4′-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.

Solubility
Soluble in chloroform and hexane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.