missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylphthalic anhydride, 96%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 414790250

 View more versions of this product

Catalog No. AC414790250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥95%
Flakes or Crystalline Powder
White to yellow-orange
100.0
Glass bottle
C9H6O3
MFCD00041856
96%
162.14
25g
88° to 93°C
95.0
Authentic
19438-61-0
4-Methylphthalic anhydride

Chemical Identifiers

19438-61-0
100.0
C9H6O3
MFCD00041856
≥95%
95.0
96%
162.14
4-Methylphthalic anhydride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.