missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylphthalic Acid, TCI America™

Manufacturer:  TCI America M0560500G

 View more versions of this product

Catalog No. M0560500G



Specifications

Specifications

4-Methylphthalic Acid
147°C
5
C9H8O4
1,2-benzenedicarboxylic acid, 4-methyl, phthalic acid, 4-methyl, 4-methyl phthalic acid, 4-methyl-phthalic acid, 4-methylbenzene-1,2-dicarboxylic acid, 4-methyl-1,2-benzenedicarboxylic acid, 4-methyl-o-phthalate, 4-methylphthalicacid, acmc-1amca, 4-methylphthalic acid
CC1=CC(=C(C=C1)C(=O)O)C(=O)O
180.159
180.16
≥98.0% (HPLC,T)
Undesignated
500g
4316-23-8
MFCD00041946
CWJJAFQCTXFSTA-UHFFFAOYSA-N
4-methylphthalic acid
20310
Crystal-Powder at 20°C

Chemical Identifiers

4316-23-8
180.159
CWJJAFQCTXFSTA-UHFFFAOYSA-N
20310
CC1=CC(=C(C=C1)C(=O)O)C(=O)O
C9H8O4
MFCD00041946
1,2-benzenedicarboxylic acid, 4-methyl, phthalic acid, 4-methyl, 4-methyl phthalic acid, 4-methyl-phthalic acid, 4-methylbenzene-1,2-dicarboxylic acid, 4-methyl-1,2-benzenedicarboxylic acid, 4-methyl-o-phthalate, 4-methylphthalicacid, acmc-1amca, 4-methylphthalic acid
4-methylphthalic acid
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1373

TSCA : Yes

Documents