Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Methylphenylthio)acetone, 97%

$55.55 - $166.24

Chemical Identifiers

Chemical Name or Material (4-Methylphenylthio)acetone
CAS 1200-13-1
Molecular Formula C10H12OS
Formula Weight 180.27
MDL Number MFCD00052713
Beilstein 2046162
Synonym p-(Tolylthio)acetone; p-(Tolylthio)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2024703

 
alfa aesar™
B2024703
1g Each for $55.55

AAB2024706

 
alfa aesar™
B2024706
5g Each for $166.24
Description & Specifications

Specifications

Formula Weight 180.27
Chemical Name or Material (4-Methylphenylthio)acetone
Assay 97%
Molecular Formula C10H12OS
MDL Number MFCD00052713
CAS 1200-13-1
Beilstein 2046162
Synonym p-(Tolylthio)acetone; p-(Tolylthio)-2-propanone
Boiling Point 96° to 98°C (0.1mmHg)
Quantity 1g