Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylphenylthio)acetaldehyde diethyl acetal, 98%

$91.91 - $374.04

Chemical Identifiers

Chemical Name or Material 4- (Methylphenylthio)acetaldehyde diethyl acetal
CAS 51830-50-3
Molecular Formula C13H20O2S
Formula Weight 240.37
MDL Number MFCD00051603
Beilstein 1964544
Synonym p-(Tolylthio)acetaldehyde diethyl acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1177006

 
alfa aesar™
A1177006
5g Each for $91.91

AAA1177014

 
alfa aesar™
A1177014
25g Each for $374.04
Description & Specifications

Specifications

Formula Weight 240.37
Chemical Name or Material 4- (Methylphenylthio)acetaldehyde diethyl acetal
Assay 98%
Molecular Formula C13H20O2S
MDL Number MFCD00051603
CAS 51830-50-3
Beilstein 1964544
Synonym p-(Tolylthio)acetaldehyde diethyl acetal
Boiling Point 106° to 108°C (0.25mmHg)
Quantity 5g