missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylphenyl beta-D-thiogalactopyranoside, 98%

$82.81 - $313.05

Chemical Identifiers

CAS 28244-98-6
Molecular Formula C13H18O5S
Molecular Weight (g/mol) 286.342
MDL Number MFCD04973631
InChI Key IQCLIQLFPVKINX-SJHCENCUSA-N
Synonym 4-methylphenylthio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyl oxane-3,4,5-triol, 4-methylphenyl b-d-thiogalactopyranoside, p-tolyl 1-thio-beta-d-galactopyranoside, 4-methylphenyl 1-thio-b-d-galactopyranoside, p-methylphenyl 1-thio-beta-d-galactopyranoside, p-methylphenyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyloxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-p-tolylthio tetrahydro-2h-pyran-3,4,5-triol
PubChem CID 7315241
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methylphenyl)sulfanyloxane-3,4,5-triol
SMILES CC1=CC=C(C=C1)SC2C(C(C(C(O2)CO)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6668603 View Documents Alfa Aesar™
J6668603
1g Each for $82.81
 
AAJ6668606 View Documents Alfa Aesar™
J6668606
5g Each for $313.05
 
Description

Description

Solubility
Soluble in water, warm methanol, DMSO, or DMF.

Notes
Store at 4°C. Protect from heat. Incompatible with heat and oxidizing agents.
Specifications

Chemical Identifiers

28244-98-6
286.342
IQCLIQLFPVKINX-SJHCENCUSA-N
7315241
CC1=CC=C(C=C1)SC2C(C(C(C(O2)CO)O)O)O
C13H18O5S
MFCD04973631
4-methylphenylthio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyl oxane-3,4,5-triol, 4-methylphenyl b-d-thiogalactopyranoside, p-tolyl 1-thio-beta-d-galactopyranoside, 4-methylphenyl 1-thio-b-d-galactopyranoside, p-methylphenyl 1-thio-beta-d-galactopyranoside, p-methylphenyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyloxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-p-tolylthio tetrahydro-2h-pyran-3,4,5-triol
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methylphenyl)sulfanyloxane-3,4,5-triol

Specifications

White
1g
C13H18O5S
4-methylphenylthio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyl oxane-3,4,5-triol, 4-methylphenyl b-d-thiogalactopyranoside, p-tolyl 1-thio-beta-d-galactopyranoside, 4-methylphenyl 1-thio-b-d-galactopyranoside, p-methylphenyl 1-thio-beta-d-galactopyranoside, p-methylphenyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyloxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-p-tolylthio tetrahydro-2h-pyran-3,4,5-triol
IQCLIQLFPVKINX-SJHCENCUSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methylphenyl)sulfanyloxane-3,4,5-triol
7315241
Powder
4-Methylphenyl beta-D-thiogalactopyranoside
137°C to 139°C
28244-98-6
MFCD04973631
Soluble in water, warm methanol, DMSO, or DMF
CC1=CC=C(C=C1)SC2C(C(C(C(O2)CO)O)O)O
286.342
286.35
98%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C