missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylnonanoic Acid, TCI America™

$67.22 - $213.89

Chemical Identifiers

CAS 45019-28-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00051789
InChI Key WQTZCQIRCYSUBQ-UHFFFAOYSA-N
Synonym nonanoic acid, 4-methyl, 4-methylpelargonic acid, 4-methyl nonanoic acid, 4-methylnonan-1-oic acid, fema no. 3574, nonanoic acid, 4-methyl-, 4r, methylpelargonic acid, acmc-20mr8v, nonanoic acid,4-methyl, acmc-1aqz1
PubChem CID 62003
IUPAC Name 4-methylnonanoic acid
SMILES CCCCCC(C)CCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M238025ML View Documents TCI America
M238025ML
25mL Each for $213.89
 
M23805ML View Documents TCI America
M23805ML
5mL Each for $67.22
 
Specifications

Chemical Identifiers

45019-28-1
172.268
WQTZCQIRCYSUBQ-UHFFFAOYSA-N
62003
CCCCCC(C)CCC(=O)O
C10H20O2
MFCD00051789
nonanoic acid, 4-methyl, 4-methylpelargonic acid, 4-methyl nonanoic acid, 4-methylnonan-1-oic acid, fema no. 3574, nonanoic acid, 4-methyl-, 4r, methylpelargonic acid, acmc-20mr8v, nonanoic acid,4-methyl, acmc-1aqz1
4-methylnonanoic acid

Specifications

145°C
25mL
C10H20O2
3265
WQTZCQIRCYSUBQ-UHFFFAOYSA-N
4-methylnonanoic acid
62003
Clear Liquid at 20°C
4-Methylnonanoic Acid
Yellow
45019-28-1
MFCD00051789
nonanoic acid, 4-methyl, 4-methylpelargonic acid, 4-methyl nonanoic acid, 4-methylnonan-1-oic acid, fema no. 3574, nonanoic acid, 4-methyl-, 4r, methylpelargonic acid, acmc-20mr8v, nonanoic acid,4-methyl, acmc-1aqz1
CCCCCC(C)CCC(=O)O
172.268
172.27
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

TSCA : Yes