Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylnicotinic acid hydrochloride, 97%

$127.18 - $522.85

Chemical Identifiers

Chemical Name or Material 4-Methylnicotinic acid hydrochloride
CAS 94015-05-1
Molecular Formula C7H7NO2·HCl
Formula Weight 173.6
MDL Number MFCD00832700
Synonym 4-Methylpyridine-3-carboxylic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2236703

  SDS
alfa aesar™
B2236703
1g Each for $127.18

AAB2236706

  SDS
alfa aesar™
B2236706
5g Each for $522.85

AAB2236701

  SDS
alfa aesar™
B2236701
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-Methylnicotinic acid hydrochloride
CAS 94015-05-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C7H7NO2·HCl
Formula Weight 173.6
MDL Number MFCD00832700
Synonym 4-Methylpyridine-3-carboxylic acid hydrochloride
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.