Learn More
4-Methylmorpholine N-oxide monohydrate, 98+%, Thermo Scientific Chemicals
$62.88 - $409.07
Chemical Identifiers
CAS | 70187-32-5 |
---|---|
Molecular Formula | C5H13NO3 |
Molecular Weight (g/mol) | 135.163 |
MDL Number | MFCD00149388 |
InChI Key | WAZPLXZGZWWXDQ-UHFFFAOYSA-N |
Synonym | 4-methylmorpholine n-oxide monohydrate, 4-methylmorpholine 4-oxide hydrate, 4-methylmorpholine n-oxide hydrate, 4-methylmorpholine 4-oxide xhydrate, unii-087wu2ftq5, n-methylmorpholine n-oxide monohydrate, n-methylmorpholine n-oxide hydrate, n-methylmorpholine-n-oxide monohydrate, 4-methyl-4-oxidomorpholin-4-ium hydrate, morpholine, 4-methyl-, 4-oxide, hydrate |
PubChem CID | 2724197 |
IUPAC Name | 4-methyl-4-oxidomorpholin-4-ium;hydrate |
SMILES | C[N+]1(CCOCC1)[O-].O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1599609
|
Thermo Scientific Chemicals
A1599609 |
10 g |
Each for $62.88
|
|
|||||
AAA1599614
|
Thermo Scientific Chemicals
A1599614 |
25 g |
Each for $126.92
|
|
|||||
AAA1599618
|
Thermo Scientific Chemicals
A1599618 |
50 g |
Each for $230.10
|
|
|||||
AAA1599622
|
Thermo Scientific Chemicals
A1599622 |
100 g |
Each for $409.07
|
|
|||||
Description
4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed. Incompatible with strong oxidizing agents.
Chemical Identifiers
70187-32-5 | |
135.163 | |
WAZPLXZGZWWXDQ-UHFFFAOYSA-N | |
2724197 | |
C[N+]1(CCOCC1)[O-].O |
C5H13NO3 | |
MFCD00149388 | |
4-methylmorpholine n-oxide monohydrate, 4-methylmorpholine 4-oxide hydrate, 4-methylmorpholine n-oxide hydrate, 4-methylmorpholine 4-oxide xhydrate, unii-087wu2ftq5, n-methylmorpholine n-oxide monohydrate, n-methylmorpholine n-oxide hydrate, n-methylmorpholine-n-oxide monohydrate, 4-methyl-4-oxidomorpholin-4-ium hydrate, morpholine, 4-methyl-, 4-oxide, hydrate | |
4-methyl-4-oxidomorpholin-4-ium;hydrate |
Specifications
70187-32-5 | |
C5H13NO3 | |
10 g | |
Hygroscopic | |
Soluble in water. | |
C[N+]1(CCOCC1)[O-].O | |
135.163 | |
135.17 (117.15 Anhydrous) | |
4-Methylmorpholine N-oxide monohydrate |
73°C to 76°C | |
MFCD00149388 | |
5449441 | |
4-methylmorpholine n-oxide monohydrate, 4-methylmorpholine 4-oxide hydrate, 4-methylmorpholine n-oxide hydrate, 4-methylmorpholine 4-oxide xhydrate, unii-087wu2ftq5, n-methylmorpholine n-oxide monohydrate, n-methylmorpholine n-oxide hydrate, n-methylmorpholine-n-oxide monohydrate, 4-methyl-4-oxidomorpholin-4-ium hydrate, morpholine, 4-methyl-, 4-oxide, hydrate | |
WAZPLXZGZWWXDQ-UHFFFAOYSA-N | |
4-methyl-4-oxidomorpholin-4-ium;hydrate | |
2724197 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 231-391-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only