Please login to your online account to display your discounted pricing

4-Methylmorpholine, 99%, Alfa Aesar™

$203.42 - $203.42

Chemical Identifiers

Chemical Name or Material 4-Methylmorpholine
CAS 109-02-4
Molecular Formula C5H11NO
Formula Weight 101.15
MDL Number MFCD00006175
UN Number UN2535
Beilstein 102719
Merck Index 14,6277
Synonym N-Methylmorpholine; NMM
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA121580F

  SDS
alfa aesar
A121580F
Each for $203.42
Description & Specifications

Specifications

Chemical Name or Material 4-Methylmorpholine
CAS 109-02-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318-H302-H312-H332;
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 4-METHYLMORPHOLINE
RTECS Number QE5775000
TSCA TSCA
Assay 99%
Molecular Formula C5H11NO
Formula Weight 101.15
MDL Number MFCD00006175
UN Number UN2535
Beilstein 102719
Merck Index 14,6277
Synonym N-Methylmorpholine; NMM
Boiling Point 116° to 118°C
Melting Point −65°C
pH 10.6
Quantity mL
Flash Point 14°C (57°F)
Odor Amine-like

Applications
4-Methylmorpholine is used as a solvent for dyes, resins, waxes and pharmaceuticals. It acts as a crosslinker in the preparation of polyurethane foams, elastomers and adhesives. It is used as a precursor to prepare N-methylmorpholine N-oxide and morpholinium cation based ionic liquids. It is utilized as corrosion inhibitors and anti-scaling agents in industries.

Solubility
Miscible with water.

Notes
Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318-H302-H312-H332;
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.