Learn More
4-Methylimidazole, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2413909
Description
4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications
4-Methylimidazole | |
49°C to 57°C | |
263°C | |
C4H6N2 | |
10 g | |
1453 | |
Soluble in water. | |
CC1=CN=CN1 | |
82.11 | |
CHEBI:40035 | |
98% |
822-36-6 | |
1.02 | |
157°C (314°F) | |
MFCD00005201 | |
UN3263 | |
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol | |
XLSZMDLNRCVEIJ-UHFFFAOYSA-N | |
5-methyl-1H-imidazole | |
13195 | |
82.11 |
Chemical Identifiers
822-36-6 | |
82.11 | |
XLSZMDLNRCVEIJ-UHFFFAOYSA-N | |
13195 | |
5-methyl-1H-imidazole |
C4H6N2 | |
MFCD00005201 | |
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol | |
CHEBI:40035 | |
CC1=CN=CN1 |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H311-H314-H335-H350
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 212-497-3
RTECSNumber : NI7350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only