missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyldiphenylamine, 98%

Manufacturer:  Alfa Aesar™ 4456009

 View more versions of this product

Catalog No. AA4456009



Specifications

Specifications

4-Methyldiphenylamine
334°C
85°C to 86°C
620-84-8
MFCD00092921
Insoluble in water. Soluble in benzene,toluene,methanol,ethanol
CC1=CC=C(C=C1)NC2=CC=CC=C2
183.254
183.25
98%
155°C (311°F)
Brown
10g
C13H13N
4-methyl-n-phenyl-aniline
AGHYMXKKEXDUTA-UHFFFAOYSA-N
4-methyl-N-phenylaniline
12109
Crystalline

Chemical Identifiers

620-84-8
183.254
AGHYMXKKEXDUTA-UHFFFAOYSA-N
12109
CC1=CC=C(C=C1)NC2=CC=CC=C2
C13H13N
MFCD00092921
4-methyl-n-phenyl-aniline
4-methyl-N-phenylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319

EINECSNumber : 210-655-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents