Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylcyclohexylamine hydrochloride, cis + trans, 98+%

$56.65 - $56.65

Chemical Identifiers

Chemical Name or Material 4-Methylcyclohexylamine hydrochloride
CAS 100959-19-1
Name Note Cis + Trans
Molecular Formula C7H15N·HCl
Formula Weight 149.67g/mol
MDL Number MFCD00051985
Beilstein 3670820
Synonym 1-Amino-4-methylcyclohexane hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0612506

 
alfa aesar™
L06125-06
5g Each for $56.65
Description & Specifications

Specifications

Melting Point ≈224°C
Quantity 5g
Chemical Name or Material 4-Methylcyclohexylamine hydrochloride
Name Note Cis + Trans
CAS 100959-19-1
Assay ≥98%
Molecular Formula C7H15N·HCl
Formula Weight 149.67g/mol
MDL Number MFCD00051985
Beilstein 3670820
Synonym 1-Amino-4-methylcyclohexane hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.