Please login to your online account to display your discounted pricing

4-Methylcyclohexylamine, 97%, mixture of cis and trans, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methylcyclohexylamine
Name Note Mixture of cis and trans
CAS 6321-23-9
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H15N
Linear Formula CH3C6H10NH2
Formula Weight 113.2
MDL Number MFCD00001495
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC175870010

  SDS
thermo fisher scientific
175870010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC175870100

  SDS
thermo fisher scientific
175870100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 113.2
Quantity 1g
Beilstein 12, 12
Density 0.85g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Methylcyclohexylamine
Name Note Mixture of cis and trans
Linear Formula CH3C6H10NH2
Assay Percent Range 97%
Molecular Formula C7H15N
CAS 6321-23-9
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00001495
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.