Please login to your online account to display your discounted pricing

4-Methylcyclohexylamine, 97%, mixture of cis and trans, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  175870100

 View more versions of this product

Catalog No. AC175870100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 113.2
Quantity 10g
Beilstein 12, 12
Density 0.85g/mL
CAS Min % 96.0
CAS Max % 100.0
Linear Formula CH3C6H10NH2
Packaging Glass bottle
Chemical Name or Material 4-Methylcyclohexylamine
Name Note Mixture of cis and trans
Assay Percent Range 97%
Molecular Formula C7H15N
CAS 6321-23-9
MDL Number MFCD00001495
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Methylcyclohexylamine
Name Note Mixture of cis and trans
CAS 6321-23-9
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H15N
Linear Formula CH3C6H10NH2
Formula Weight 113.2
MDL Number MFCD00001495
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents