missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylcyclohexanone, 98%, ACROS Organics™

Manufacturer:  Acros Organics 174260250

 View more versions of this product

Catalog No. AC174260250



Specifications

Specifications

4-Methylcyclohexanone
Authentic
1.4445 to 1.4465 (20°C, 589nm)
25g
589-92-4
100.0
C7H12O
MFCD00001643
VGVHNLRUAMRIEW-UHFFFAOYSA-N
4-methylcyclohexan-1-one
11525
Liquid
0.9140g/mL
Glass bottle
Colorless to Yellow
98%
97.5
98%
CH3C6H9(=O)
4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french
CC1CCC(=O)CC1
112.172
112.17
≥97.5%

Chemical Identifiers

589-92-4
112.172
VGVHNLRUAMRIEW-UHFFFAOYSA-N
11525
CC1CCC(=O)CC1
C7H12O
MFCD00001643
4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french
4-methylcyclohexan-1-one
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-665-3

RTECSNumber : GW1750200

TSCA : TSCA

SDS Documents