missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylcyclohexanol, 99%, mixture of cis and trans, ACROS Organics™

Manufacturer:  Acros Organics 166900050

 View more versions of this product

Catalog No. AC166900050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methylcyclohexanol
68°C
Glass bottle
0.92
Colorless to Yellow
5g
589-91-3
100.0
C7H14O
MFCD00001449
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
CC1CCC(CC1)O
114.188
114.19
99%
0.9200g/mL
Authentic
1.4570 to 1.4590
171.0°C to 173.0°C
-41.0°C
99%
98.5
99%
CH3C6H10OH
06, 14
MQWCXKGKQLNYQG-UHFFFAOYSA-N
4-methylcyclohexan-1-ol
11524
Viscous Liquid

Chemical Identifiers

589-91-3
114.188
MQWCXKGKQLNYQG-UHFFFAOYSA-N
11524
CC1CCC(CC1)O
C7H14O
MFCD00001449
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
4-methylcyclohexan-1-ol
Safety and Handling

Safety and Handling

GHS P Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 209-664-8

TSCA : TSCA

SDS Documents