missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylcyclohexane-1,2-dicarboxylic Anhydride, TCI America™

Manufacturer:  TCI America M056725G

 View more versions of this product

Catalog No. M056725G



Specifications

Specifications

4-Methylcyclohexane-1,2-dicarboxylic Anhydride
25g
C9H12O3
hexahydro-4-methylphthalic anhydride, 4-methylhexahydrophthalic anhydride, 5-methylhexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, 1,3-isobenzofurandione, hexahydro-5-methyl, 1,3-isobenzofurandione, hexahydromethyl, 4-methylcyclohexane-1,2-dicarboxylic anhydride, 5-methylhexahydrophthalic anhydride, 5-methyl-3a,4,5,6,7,7a-hexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers
CC1CCC2C(C1)C(=O)OC2=O
168.192
168.19
≥98.0% (GC)
Colorless
19438-60-9
MFCD00005927
FKBMTBAXDISZGN-UHFFFAOYSA-N
5-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
86876
Clear Liquid at 20°C

Chemical Identifiers

19438-60-9
168.192
FKBMTBAXDISZGN-UHFFFAOYSA-N
86876
CC1CCC2C(C1)C(=O)OC2=O
C9H12O3
MFCD00005927
hexahydro-4-methylphthalic anhydride, 4-methylhexahydrophthalic anhydride, 5-methylhexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, 1,3-isobenzofurandione, hexahydro-5-methyl, 1,3-isobenzofurandione, hexahydromethyl, 4-methylcyclohexane-1,2-dicarboxylic anhydride, 5-methylhexahydrophthalic anhydride, 5-methyl-3a,4,5,6,7,7a-hexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers
5-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2450

TSCA : Yes

Documents