Please login to your online account to display your discounted pricing

4-Methylcinnamic acid, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methylcinnamic acid
Name Note Predominantly trans
CAS 1866-39-3
Assay ≥98.5%
Molecular Formula C10H10O2
Linear Formula CH3C6H4CH=CHCO2H
Formula Weight 162.19
MDL Number MFCD00002697
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126560050

  SDS
thermo fisher scientific
126560050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 162.19
Quantity 5g
Color White
Beilstein 09, 617
Melting Point 198° to 201°C
Linear Formula CH3C6H4CH=CHCO2H
CAS 1866-39-3
Chemical Name or Material 4-Methylcinnamic acid
Infrared Spectrum Authentic
Name Note Predominantly trans
MDL Number MFCD00002697
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Molecular Formula C10H10O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.