Please login to your online account to display your discounted pricing

4-Methylcinnamic acid, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  126561000

 View more versions of this product

Catalog No. AC126561000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 198° to 201°C
Color White
Quantity 100g
Chemical Name or Material 4-Methylcinnamic acid
Name Note Predominantly trans
CAS 1866-39-3
Physical Form White fine crystalline powder
Assay 98.5% min.
Molecular Formula C10H10O2
Linear Formula CH3C6H4CH=CHCO2H
Formula Weight 162.19g/mol
MDL Number MFCD00002697
Beilstein 09, 617
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Methylcinnamic acid
Name Note Predominantly trans
CAS 1866-39-3
Assay 98.5% min.
Molecular Formula C10H10O2
Linear Formula CH3C6H4CH=CHCO2H
Formula Weight 162.19g/mol
MDL Number MFCD00002697
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents