missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylcatechol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC126515000

 View more versions of this product

Catalog No. AC126515000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

124.14
≥97.5%
White to brown
06, 878
CH3C6H3(OH)2
452-86-8
97.5
3, 4-Dihydroxytoluene
98%
Powder or Chunks
500g
63° to 68°C
100.0
C7H8O2
4-Methylcatechol
MFCD00002205
Glass bottle
Authentic

Chemical Identifiers

452-86-8
100.0
C7H8O2
124.14
3, 4-Dihydroxytoluene
≥97.5%
97.5
98%
CH3C6H3(OH)2
MFCD00002205
4-Methylcatechol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning