missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylcarbamoyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53090MD

 View more versions of this product

Catalog No. AAH53090MD



Specifications

Specifications

4-(Methylcarbamoyl)benzeneboronic acid
250mg
C8H10BNO3
4-n-methylaminocarbonyl phenylboronic acid, 4-methylcarbamoyl phenylboronic acid, 4-methylcarbamoyl phenyl boronic acid, 4-methylcarbamoyl benzeneboronic acid, 4-methylcarbamoyl phenyl boronic acid, boronic acid, 4-methylamino carbonyl phenyl, boronic acid,b-4-methylamino carbonyl phenyl, 4-methylamino carbonyl phenylboronic acid, 4-n-methylaminocarbonyl benzeneboronic acid, zlchem 836
B(C1=CC=C(C=C1)C(=O)NC)(O)O
178.982
178.98
166°C to 168°C
121177-82-0
MFCD03788427
UWKSYZHFTGONHY-UHFFFAOYSA-N
[4-(methylcarbamoyl)phenyl]boronic acid
2773526
98%

Chemical Identifiers

121177-82-0
178.982
UWKSYZHFTGONHY-UHFFFAOYSA-N
2773526
B(C1=CC=C(C=C1)C(=O)NC)(O)O
C8H10BNO3
MFCD03788427
4-n-methylaminocarbonyl phenylboronic acid, 4-methylcarbamoyl phenylboronic acid, 4-methylcarbamoyl phenyl boronic acid, 4-methylcarbamoyl benzeneboronic acid, 4-methylcarbamoyl phenyl boronic acid, boronic acid, 4-methylamino carbonyl phenyl, boronic acid,b-4-methylamino carbonyl phenyl, 4-methylamino carbonyl phenylboronic acid, 4-n-methylaminocarbonyl benzeneboronic acid, zlchem 836
[4-(methylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents