missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylbiphenyl, 98%

Manufacturer:  Alfa Aesar™ A1470203

 View more versions of this product

Catalog No. AAA1470203



Description

Description

Applications
4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

Solubility
Insoluble in water. Soluble in alcohol and ether, very soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

4-Methylbiphenyl
267°C to 268°C
1g
C13H12
1904468
ZZLCFHIKESPLTH-UHFFFAOYSA-N
1-methyl-4-phenylbenzene
12566
98%
>110°C (230°F)
44°C to 48°C
644-08-6
MFCD00008544
4-methylbiphenyl, 4-phenyltoluene, 4-methyl-1,1'-biphenyl, p-methylbiphenyl, p-methyldiphenyl, 4-methyldiphenyl, 1,1'-biphenyl, 4-methyl, biphenyl, 4-methyl, p-phenyltoluene, 4-methylphenyl benzene
CC1=CC=C(C=C1)C2=CC=CC=C2
168.239
168.24

Chemical Identifiers

644-08-6
168.239
ZZLCFHIKESPLTH-UHFFFAOYSA-N
12566
CC1=CC=C(C=C1)C2=CC=CC=C2
C13H12
MFCD00008544
4-methylbiphenyl, 4-phenyltoluene, 4-methyl-1,1'-biphenyl, p-methylbiphenyl, p-methyldiphenyl, 4-methyldiphenyl, 1,1'-biphenyl, 4-methyl, biphenyl, 4-methyl, p-phenyltoluene, 4-methylphenyl benzene
1-methyl-4-phenylbenzene
Safety and Handling

Safety and Handling

RTECSNumber : DV5460000

TSCA : TSCA

Documents