4'-Methylbiphenyl-4-sulfonyl chloride, 97%, Acros Organics™
Manufacturer:  Acros Organics AC450410050
 View more versions of this product
Catalog No. AC450410050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
4'-Methylbiphenyl-4-sulfonyl chloride
107.0°C to 110.0°C
101366-51-2
4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl
CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
266.739
266.75
Authentic
5g
C13H11ClO2S
DBYXACYHGMSPMQ-UHFFFAOYSA-N
4-(4-methylphenyl)benzenesulfonyl chloride
2794758
97%
Chemical Identifiers
101366-51-2
266.739
4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl
4-(4-methylphenyl)benzenesulfonyl chloride
C13H11ClO2S
DBYXACYHGMSPMQ-UHFFFAOYSA-N
2794758
CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Safety and Handling
Safety and Handling
GHS H Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water.
GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container.
Danger
Documents