Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Methylbenzyl)triphenylphosphonium chloride, 98+%

$55.55 - $195.49

Chemical Identifiers

Chemical Name or Material (4-Methylbenzyl)triphenylphosphonium chloride
CAS 1530-37-6
Molecular Formula C26H24ClP
Formula Weight 402.91
MDL Number MFCD00031581
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1244709

 
alfa aesar™
A1244709
10g Each for $55.55

AAA1244718

 
alfa aesar™
A1244718
50g Each for $195.49
Description & Specifications

Specifications

Formula Weight 402.91
Chemical Name or Material (4-Methylbenzyl)triphenylphosphonium chloride
Assay ≥98%
Molecular Formula C26H24ClP
MDL Number MFCD00031581
CAS 1530-37-6
Health Hazard 1 Warning
Melting Point 260° to 262°C
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.