missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Methylbenzyl)triphenylphosphonium bromide, 98+%

$55.51 - $192.77

Chemical Identifiers

CAS 2378-86-1
Molecular Formula C26H24BrP
Molecular Weight (g/mol) 447.356
MDL Number MFCD00031580
InChI Key CEEKCHGNKYVTTM-UHFFFAOYSA-M
Synonym 4-methylbenzyl triphenylphosphonium bromide, 4-methylphenyl methyl triphenylphosphanium bromide, pubchem2184, acmc-1cfpa, 4-methylphenyl methyl-triphenyl-phosphanium bromide, 4-methylbenzyl triphenylphosphoniumbromide, 4-methylbenzyl triphenylphosphonium bromide, 4-methylphenyl methyl-triphenylphosphonium bromide, p-methylbenzyl-triphenylphosphomiumbromide, phosphonium, 4-methylphenyl methyl triphenyl-, bromide 1:1
PubChem CID 11797662
IUPAC Name (4-methylphenyl)methyl-triphenylphosphanium;bromide
SMILES CC1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0156206 View Documents Alfa Aesar™
L0156206
5g Each for $55.51
 
AAL0156214 View Documents Alfa Aesar™
L0156214
25g Each for $192.77
 
Specifications

Chemical Identifiers

2378-86-1
447.356
CEEKCHGNKYVTTM-UHFFFAOYSA-M
11797662
CC1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
C26H24BrP
MFCD00031580
4-methylbenzyl triphenylphosphonium bromide, 4-methylphenyl methyl triphenylphosphanium bromide, pubchem2184, acmc-1cfpa, 4-methylphenyl methyl-triphenyl-phosphanium bromide, 4-methylbenzyl triphenylphosphoniumbromide, 4-methylbenzyl triphenylphosphonium bromide, 4-methylphenyl methyl-triphenylphosphonium bromide, p-methylbenzyl-triphenylphosphomiumbromide, phosphonium, 4-methylphenyl methyl triphenyl-, bromide 1:1
(4-methylphenyl)methyl-triphenylphosphanium;bromide

Specifications

268°C to 270°C
2378-86-1
MFCD00031580
4-methylbenzyl triphenylphosphonium bromide, 4-methylphenyl methyl triphenylphosphanium bromide, pubchem2184, acmc-1cfpa, 4-methylphenyl methyl-triphenyl-phosphanium bromide, 4-methylbenzyl triphenylphosphoniumbromide, 4-methylbenzyl triphenylphosphonium bromide, 4-methylphenyl methyl-triphenylphosphonium bromide, p-methylbenzyl-triphenylphosphomiumbromide, phosphonium, 4-methylphenyl methyl triphenyl-, bromide 1:1
CC1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
447.356
447.36
(4-Methylbenzyl)triphenylphosphonium bromide
5g
C26H24BrP
3810292
CEEKCHGNKYVTTM-UHFFFAOYSA-M
(4-methylphenyl)methyl-triphenylphosphanium;bromide
11797662
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning