missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylbenzyl Mercaptan, TCI America™

Manufacturer:  TCI America M068210ML

 View more versions of this product

Catalog No. M068210ML



Specifications

Specifications

4-Methylbenzyl Mercaptan
Yellow
4498-99-1
MFCD00039652
AGFYZLVFPSGUIX-UHFFFAOYSA-N
(4-methylphenyl)methanethiol
78252
Clear Liquid at 20°C
118°C
10mL
C8H10S
4-methylbenzyl mercaptan, 4-methylphenyl methanethiol, p-xylene-alpha-thiol, alpha-mercapto-p-xylene, 4-methylbenzenemethanethiol, 4-methylbenzylmercaptan, benzenemethanethiol, 4-methyl, p-xylyl mercaptan, 4-methylphenyl methane-1-thiol, acmc-1aqt2
CC1=CC=C(C=C1)CS
138.228
138.23
≥97.0% (GC)

Chemical Identifiers

4498-99-1
138.228
AGFYZLVFPSGUIX-UHFFFAOYSA-N
78252
CC1=CC=C(C=C1)CS
C8H10S
MFCD00039652
4-methylbenzyl mercaptan, 4-methylphenyl methanethiol, p-xylene-alpha-thiol, alpha-mercapto-p-xylene, 4-methylbenzenemethanethiol, 4-methylbenzylmercaptan, benzenemethanethiol, 4-methyl, p-xylyl mercaptan, 4-methylphenyl methane-1-thiol, acmc-1aqt2
(4-methylphenyl)methanethiol
Safety and Handling

Safety and Handling

TSCA : No

Documents