missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylbenzyl Mercaptan, TCI America™

$302.50 - $511.81

Chemical Identifiers

CAS 4498-99-1
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00039652
InChI Key AGFYZLVFPSGUIX-UHFFFAOYSA-N
Synonym 4-methylbenzyl mercaptan, 4-methylphenyl methanethiol, p-xylene-alpha-thiol, alpha-mercapto-p-xylene, 4-methylbenzenemethanethiol, 4-methylbenzylmercaptan, benzenemethanethiol, 4-methyl, p-xylyl mercaptan, 4-methylphenyl methane-1-thiol, acmc-1aqt2
PubChem CID 78252
IUPAC Name (4-methylphenyl)methanethiol
SMILES CC1=CC=C(C=C1)CS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M068210ML View Documents TCI America
M068210ML
10mL Each for $302.50
 
M068225ML View Documents TCI America
M068225ML
25mL Each for $511.81
 
Specifications

Chemical Identifiers

4498-99-1
138.228
AGFYZLVFPSGUIX-UHFFFAOYSA-N
78252
CC1=CC=C(C=C1)CS
C8H10S
MFCD00039652
4-methylbenzyl mercaptan, 4-methylphenyl methanethiol, p-xylene-alpha-thiol, alpha-mercapto-p-xylene, 4-methylbenzenemethanethiol, 4-methylbenzylmercaptan, benzenemethanethiol, 4-methyl, p-xylyl mercaptan, 4-methylphenyl methane-1-thiol, acmc-1aqt2
(4-methylphenyl)methanethiol

Specifications

118°C
10mL
C8H10S
4-methylbenzyl mercaptan, 4-methylphenyl methanethiol, p-xylene-alpha-thiol, alpha-mercapto-p-xylene, 4-methylbenzenemethanethiol, 4-methylbenzylmercaptan, benzenemethanethiol, 4-methyl, p-xylyl mercaptan, 4-methylphenyl methane-1-thiol, acmc-1aqt2
CC1=CC=C(C=C1)CS
138.228
138.23
≥97.0% (GC)
Yellow
4498-99-1
MFCD00039652
AGFYZLVFPSGUIX-UHFFFAOYSA-N
(4-methylphenyl)methanethiol
78252
Clear Liquid at 20°C
4-Methylbenzyl Mercaptan
Safety and Handling

Safety and Handling

TSCA : No