Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylbenzyl mercaptan, 97%

Chemical Identifiers

Chemical Name or Material 4-Methylbenzyl mercaptan
CAS 4498-99-1
Molecular Formula C8H10S
Formula Weight 138.23
MDL Number MFCD00039652
UN Number UN2810
Beilstein 2411317
Synonym 4-Methyl-alpha-toluenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1046706

  SDS
alfa aesar™
A1046706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1046714

  SDS
alfa aesar™
A1046714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1046722

  SDS
alfa aesar™
A1046722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.23
Chemical Name or Material 4-Methylbenzyl mercaptan
Assay 97%
Molecular Formula C8H10S
MDL Number MFCD00039652
CAS 4498-99-1
UN Number UN2810
Beilstein 2411317
Synonym 4-Methyl-alpha-toluenethiol
Health Hazard 1 Danger
Boiling Point 116° to 118°C (35mmHg)
Quantity 5g
Odor Stench
Health Hazard 2 GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Applications
4-Methylbenzyl mercaptan is used as pharmaceutical intermediates.

Solubility
Sparingly Soluble.

Notes
Air Sensitive. Store away from air and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.