Learn More
4-Methylbenzyl bromide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1329706
Description
4-Methylbenzyl bromide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methylbenzyl bromide is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Specifications
4-Methylbenzyl bromide | |
33°C to 36°C | |
218°C to 220°C | |
C8H9Br | |
5 g | |
507389 | |
4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene | |
WZRKSPFYXUXINF-UHFFFAOYSA-N | |
1-(bromomethyl)-4-methylbenzene | |
7721 | |
98% |
104-81-4 | |
1.324 | |
97°C (206°F) | |
MFCD00000181 | |
UN3417 | |
14,10090 | |
Insoluble in water. | |
CC1=CC=C(C=C1)CBr | |
185.064 | |
185.07 |
Chemical Identifiers
104-81-4 | |
185.064 | |
WZRKSPFYXUXINF-UHFFFAOYSA-N | |
7721 | |
CC1=CC=C(C=C1)CBr |
C8H9Br | |
MFCD00000181 | |
4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene | |
1-(bromomethyl)-4-methylbenzene |
Safety and Handling
GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLYL BROMIDE, SOLID
EINECSNumber : 203-240-6
RTECSNumber : CY9039300
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only