Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylbenzyl acetate, 98+%

$543.36 - $543.36

Chemical Identifiers

Chemical Name or Material 4-Methylbenzyl acetate
CAS 2216-45-7
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00017234
Beilstein 2046163
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2231830

 
alfa aesar™
B2231830
250g Each for $543.36
Description & Specifications

Specifications

Formula Weight 164.2
Chemical Name or Material 4-Methylbenzyl acetate
TSCA TSCA
Assay ≥98%
Molecular Formula C10H12O2
MDL Number MFCD00017234
CAS 2216-45-7
Beilstein 2046163
Boiling Point ≈100°C (10mmHg)
Quantity 250g
RTECS Number DA4940000