Please login to your online account to display your discounted pricing

4-Methylbenzoylacetonitrile, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methylbenzoylacetonitrile
CAS 7391-28-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H9NO
Formula Weight 159.19
MDL Number MFCD00067895
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC355060010

  SDS
thermo fisher scientific
35506-0010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Needle-Like Powder
Formula Weight 159.19
Quantity 1g
Color Light beige
Melting Point 104° to 108°C
CAS Max % 100.0
Chemical Name or Material 4-Methylbenzoylacetonitrile
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C10H9NO
MDL Number MFCD00067895
CAS 7391-28-8
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.