missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylbenzotrifluoride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 308912500

 View more versions of this product

Catalog No. AC308912500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
160.14
Clear colorless
1.144g/mL
MFCD00075476
1.4250 to 1.4270 (20°C, 589nm)
Authentic
98%
C8H7F3
CH3C6H4CF3
Liquid
250mg
Glass Bottle
100.0
97.5
α, α, α-Trifluoro-p-xylene
4-Methylbenzotrifluoride
98%
6140-17-6

Chemical Identifiers

6140-17-6
100.0
C8H7F3
160.14
α, α, α-Trifluoro-p-xylene
≥97.5%
97.5
98%
CH3C6H4CF3
MFCD00075476
4-Methylbenzotrifluoride, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-124-2