Please login to your online account to display your discounted pricing

4-Methylbenzotrifluoride, 98%, ACROS Organics™

$56.20 - $229.86

Chemical Identifiers

Chemical Name or Material 4-Methylbenzotrifluoride
Name Note 98%
CAS 6140-17-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7F3
Linear Formula CH3C6H4CF3
Formula Weight 160.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC308910050

  SDS
thermo fisher scientific
308910050
Glass bottle 5g Each for $229.86

AC308912500

  SDS
thermo fisher scientific
308912500
Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC308910010

  SDS
thermo fisher scientific
AC308910010
Glass bottle 1g Each for $56.20
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 160.14
Quantity 5g
Color Clear colorless
EINECS Number 228-124-2
Density 1.144g/mL
CAS Max % 100.0
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Synonym α, α, α-Trifluoro-p-xylene
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Methylbenzotrifluoride
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H7F3
Linear Formula CH3C6H4CF3
MDL Number MFCD00075476
CAS 6140-17-6
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.