missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylbenzophenone, 98%

Manufacturer:  Alfa Aesar™ A1547918

 View more versions of this product

Catalog No. AAA1547918



Description

Description

Applications
4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place. Store in a tightly closed container. Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Methylbenzophenone
326°C
50g
C14H12O
1909310
4-methylbenzophenone, phenyl p-tolyl methanone, 4-methylphenyl phenyl methanone, p-methylbenzophenone, methanone, 4-methylphenyl phenyl, p-benzoyltoluene, 4-methyl benzophenone, phenyl p-tolyl ketone, benzophenone, 4-methyl, p-benzophenone, methyl
CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
196.249
196.25
143°C (289°F)
53°C to 57°C
134-84-9
MFCD00008553
14,7317
WXPWZZHELZEVPO-UHFFFAOYSA-N
(4-methylphenyl)-phenylmethanone
8652
98%

Chemical Identifiers

134-84-9
196.249
WXPWZZHELZEVPO-UHFFFAOYSA-N
8652
CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
C14H12O
MFCD00008553
4-methylbenzophenone, phenyl p-tolyl methanone, 4-methylphenyl phenyl methanone, p-methylbenzophenone, methanone, 4-methylphenyl phenyl, p-benzoyltoluene, 4-methyl benzophenone, phenyl p-tolyl ketone, benzophenone, 4-methyl, p-benzophenone, methyl
(4-methylphenyl)-phenylmethanone
Safety and Handling

Safety and Handling

RTECSNumber : DJ1750000

TSCA : TSCA

Documents