missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylbenzhydrylamine hydrochloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-1.0 mmol/g on PS-DVB

Manufacturer:  Alfa Aesar™ 4445514

 View more versions of this product

Catalog No. AA4445514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methylbenzhydrylamine hydrochloride
White to Yellow
Polymer-supported, 1% cross-linked, 0.5 to 1.0 mmol/g on PS-DVB
C14H16ClN
4-methylbenzhydrylamine hydrochloride
NHTZSJKMWBONMD-UHFFFAOYNA-N
2734294
100 to 200 Mesh
25g
5267-49-2
MFCD00801150
Suspended in DCM/DFM(1:1).
233.74
Beads

Chemical Identifiers

5267-49-2
233.74
NHTZSJKMWBONMD-UHFFFAOYNA-N
2734294
C14H16ClN
MFCD00801150
4-methylbenzhydrylamine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents