Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylbenzhydrol, 98+%

$26.00 - $585.80

Chemical Identifiers

Chemical Name or Material 4-Methylbenzhydrol
CAS 1517-63-1
Molecular Formula C14H14O
Formula Weight 198.27g/mol
MDL Number MFCD00004492
Beilstein 2048849
Synonym 4-Methyldiphenylmethanol; Phenyl p-tolyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1690414

 
alfa aesar™
A16904-14
25g Each for $26.00

AAA1690436

 
alfa aesar™
A16904-36
500g Each for $585.80

AAA1690422

 
alfa aesar™
A16904-22
100g Each for $94.54
Description & Specifications

Specifications

Melting Point 50° to 54°C
Quantity 25g
Chemical Name or Material 4-Methylbenzhydrol
CAS 1517-63-1
Assay ≥98%
Molecular Formula C14H14O
Formula Weight 198.27g/mol
MDL Number MFCD00004492
Beilstein 2048849
Synonym 4-Methyldiphenylmethanol; Phenyl p-tolyl carbinol
TSCA TSCA