Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methylbenzenethiol sodium salt, 98%

Chemical Identifiers

Chemical Name or Material 4-Methylbenzenethiol sodium salt
CAS 10486-08-5
Formula Weight 146.18
MDL Number MFCD00066208
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5509503

  SDS
alfa aesar™
H5509503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5509506

  SDS
alfa aesar™
H5509506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5509514

  SDS
alfa aesar™
H5509514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.18
Chemical Name or Material 4-Methylbenzenethiol sodium salt
Physical Form Powder
Assay 98%
MDL Number MFCD00066208
CAS 10486-08-5
Color White to off-white
Melting Point >300°C
Quantity 1g

Applications
4-Methylbenzenethiol sodium salt is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.