Learn More
4-Methylbenzenesulfonhydrazide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 157865000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methylbenzenesulfonhydrazide | |
96.0 | |
109.0°C to 113.0°C | |
Authentic | |
Plastic bottle | |
CH3C6H4SO2NHNH2 | |
500 g | |
01,1185; 02,417; 03,293; 04,511; 05,678; 06,598; 07,375; 08,489; 09,472 | |
4-methylbenzenesulfonhydrazide, p-toluenesulfonhydrazide, tosylhydrazine, p-toluenesulfonyl hydrazide, tosylhydrazide, p-tosylhydrazine, p-toluenesulfonylhydrazine, celogen tsh, ptsh, 4-toluenesulfonyl hydrazide | |
ICGLPKIVTVWCFT-UHFFFAOYSA-N | |
4-methylbenzenesulfonohydrazide | |
15303 | |
97% |
1576-35-8 | |
100.0 | |
Gray to White or White | |
96% min. (HPLC) | |
C7H10N2O2S | |
MFCD00007588 | |
11, II, 66 | |
15, 9694 | |
Solubility in water: 5g/L (15°C) | |
CC1=CC=C(C=C1)S(=O)(=O)NN | |
186.23 | |
186.23 | |
Powder |
Chemical Identifiers
1576-35-8 | |
186.23 | |
ICGLPKIVTVWCFT-UHFFFAOYSA-N | |
15303 |
C7H10N2O2S | |
MFCD00007588 | |
4-methylbenzenesulfonhydrazide, p-toluenesulfonhydrazide, tosylhydrazine, p-toluenesulfonyl hydrazide, tosylhydrazide, p-tosylhydrazine, p-toluenesulfonylhydrazine, celogen tsh, ptsh, 4-toluenesulfonyl hydrazide | |
CC1=CC=C(C=C1)S(=O)(=O)NN |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Heating may cause a fire.
Causes serious eye irritation.
Risk of explosion if heated under confinement.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
Keep only in orig
GHS Signal Word: Danger
EINECSNumber : 216-407-3
RUO – Research Use Only