missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylbenzenesulfonhydrazide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 157860250

 View more versions of this product

Catalog No. AC157860250



Specifications

Specifications

4-Methylbenzenesulfonhydrazide
Plastic bottle
109.0°C to 113.0°C
1576-35-8
100.0
C7H10N2O2S
MFCD00007588
01,1185; 02,417; 03,293; 04,511; 05,678; 06,598; 07,375; 08,489; 09,472
4-methylbenzenesulfonhydrazide
ICGLPKIVTVWCFT-UHFFFAOYSA-N
4-methylbenzenesulfonohydrazide
15303
Powder
Authentic
Gray to White or White
25g
96.0
96% min. (HPLC)
CH3C6H4SO2NHNH2
11, II, 66
15, 9694
Solubility in water: 5g/L (15°C)
CC1=CC=C(C=C1)S(=O)(=O)NN
186.23
186.23
97%

Chemical Identifiers

1576-35-8
186.23
ICGLPKIVTVWCFT-UHFFFAOYSA-N
15303
CC1=CC=C(C=C1)S(=O)(=O)NN
C7H10N2O2S
MFCD00007588
4-methylbenzenesulfonhydrazide
4-methylbenzenesulfonohydrazide
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Heating may cause a fire.
Causes serious eye irritation.
Risk of explosion if heated under confinement.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
Keep only in orig

GHS Signal Word: Danger

EINECSNumber : 216-407-3

SDS Documents