missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1334706

 View more versions of this product

Catalog No. AAA1334706



Description

Description

4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

4-Methylbenzeneboronic acid
246°C to 254°C
MFCD00039138
2935970
It is soluble in water.
CC1=CC=C(C=C1)B(O)O
135.96
135.96
5720-05-8
C7H9BO2
5 g
4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
BIWQNIMLAISTBV-UHFFFAOYSA-N
(4-methylphenyl)boronic acid
79799
98%

Chemical Identifiers

5720-05-8
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : XS7400000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only