Learn More
4-Methylaminophenol sulfate, 99%, extra pure, Thermo Scientific™
4-Methylaminophenol sulfate, 99%, C14H18N2O2.H2SO4, CAS Number-55-55-0, Metol, 500g, 13, 441, 100.0, 98.5, Beige to White or Gray, Decomposition Information: >260 deg.C, 1.5770g/mL, 200-237-1, 344.38, Extra Pure, GHS Signal Word: Warning
Supplier: Thermo Scientific Chemicals 156800025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Methylaminophenol sulfate | |
55-55-0 | |
100.0 | |
1.5770g/mL | |
0.3% max. | |
99% | |
C14H18N2O2·H2SO4 | |
MFCD00013140 | |
13, 441 | |
15, 6091 | |
Solubility in water: 50g/L (20°C). Other solubilities: soluble in 20 parts cold water,soluble in 6 parts boiling water,slightly soluble in alcohol,insoluble in ether | |
344.38 | |
Extra Pure |
99% | |
98.5 | |
Decomposition Information: >260°C | |
0.15% max. | |
Authentic | |
Plastic bottle | |
(CH3NHC6H4OH)2·H2SO4 | |
2.5kg | |
1.577 | |
Metol | |
344.38 | |
99% |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation . May cause an allergic skin reaction. Danger of serious damage to health by prolonged exposure if swallowed. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health: 2
Flammability: 1
Instability: 1
GHS Signal Word: Warning
EINECSNumber : 200-237-1
RTECSNumber : SL8650000
TSCA : TSCA
RUO â Research Use Only