missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methylamino) Phenol Sulfate Solution, Certified, LabChem™

$83.94 - $83.94

Chemical Identifiers

CAS 7732-18-5,7631-90-5,55-55-0
CAS Max % 79.8,20,0.2
Linear Formula 2(C7H9NO)·H2SO4
Formula Weight 344.38
Chemical Name or Material 4-(Methylamino) Phenol Sulfate Solution
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC168401 Lab Chem
LC168401
500mL Each for $83.94
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7631-90-5,55-55-0
2(C7H9NO)·H2SO4
4-(Methylamino) Phenol Sulfate Solution
79.8,20,0.2
344.38
Certified Grade

Specifications

7732-18-5,7631-90-5,55-55-0
Liquid
344.38
Poly Bottle
Passes Test
Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
500mL
Certified Grade
79.8,20,0.2
2(C7H9NO)·H2SO4
Passes Test
Soluble in water
Room Temperature
Clear yellow
4-(Methylamino) Phenol Sulfate Solution

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Contaminated work clothing must not be allowed out of the workplace.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.

Warning