Please login to your online account to display your discounted pricing

4-(Methylamino)phenol Sulfate RS, ACS Reagent Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material 4-(Methylamino)phenol Sulfate RS
Purity Grade ACS Reagent Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

4839-16

 
ricca chemical company
RI483916
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

4839-4

 
ricca chemical company
RI48394
Glass 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Quantity 500mL
Chemical Name or Material 4-(Methylamino)phenol Sulfate RS
Packaging Glass