Learn More
4-(Methylamino)benzonitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6514806
Description
4-(Methylamino)benzonitrile is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Methylamino)benzonitrile is used as pharmaceutical intermediates.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
4-(Methylamino)benzonitrile | |
87°C to 91°C | |
MFCD00016381 | |
2802435 | |
Soluble in methanol. | |
CNC1=CC=C(C=C1)C#N | |
132.166 | |
132.17 |
4714-62-9 | |
C8H8N2 | |
5 g | |
4-methylamino benzonitrile, 4-n-methylamino benzonitrile, 4-cyano-n-methylaniline, 4-methylamino-benzonitrile, benzonitrile, 4-methylamino, pubchem4815, 4-cyano-n-methyl-aniline, ksc497o2f, 4-n-methyl aminobenzonitrile, 4-n-methylamino-benzonitrile | |
JEDXDYXIRZOEKG-UHFFFAOYSA-N | |
4-(methylamino)benzonitrile | |
10964551 | |
97% |
Chemical Identifiers
4714-62-9 | |
132.166 | |
JEDXDYXIRZOEKG-UHFFFAOYSA-N | |
10964551 | |
CNC1=CC=C(C=C1)C#N |
C8H8N2 | |
MFCD00016381 | |
4-methylamino benzonitrile, 4-n-methylamino benzonitrile, 4-cyano-n-methylaniline, 4-methylamino-benzonitrile, benzonitrile, 4-methylamino, pubchem4815, 4-cyano-n-methyl-aniline, ksc497o2f, 4-n-methyl aminobenzonitrile, 4-n-methylamino-benzonitrile | |
4-(methylamino)benzonitrile |
Safety and Handling
GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only