missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methylamino)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H6514814

 View more versions of this product

Catalog No. AAH6514814



Description

Description

Applications
4-(Methylamino)benzonitrile is used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

4-(Methylamino)benzonitrile
25g
C8H8N2
2802435
JEDXDYXIRZOEKG-UHFFFAOYSA-N
4-(methylamino)benzonitrile
10964551
97%
87°C to 91°C
4714-62-9
MFCD00016381
4-methylamino benzonitrile, 4-n-methylamino benzonitrile, 4-cyano-n-methylaniline, 4-methylamino-benzonitrile, benzonitrile, 4-methylamino, pubchem4815, 4-cyano-n-methyl-aniline, ksc497o2f, 4-n-methyl aminobenzonitrile, 4-n-methylamino-benzonitrile
CNC1=CC=C(C=C1)C#N
132.166
132.17

Chemical Identifiers

4714-62-9
132.166
JEDXDYXIRZOEKG-UHFFFAOYSA-N
10964551
CNC1=CC=C(C=C1)C#N
C8H8N2
MFCD00016381
4-methylamino benzonitrile, 4-n-methylamino benzonitrile, 4-cyano-n-methylaniline, 4-methylamino-benzonitrile, benzonitrile, 4-methylamino, pubchem4815, 4-cyano-n-methyl-aniline, ksc497o2f, 4-n-methyl aminobenzonitrile, 4-n-methylamino-benzonitrile
4-(methylamino)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents